1-Bromo-4-chloro-2-pyrrolidinobenzene 96%
Dodavatel: Apollo Scientific
|
Varování
|
OR900017-10G
OR900017-1G
OR900017-5G
APOSOR900017-10GEA
15050
CZK
APOSOR900017-10G
APOSOR900017-1G
APOSOR900017-5G
1-Bromo-4-chloro-2-pyrrolidinobenzene 96%
1-Bromo-4-chloro-2-pyrrolidinobenzene
Vzorec:
C₁₀H₁₁BrClN MW: 260,558 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD18087713 CAS číslo: 1257664-93-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...