Tisk…

3-(Bromomethyl)phenyl acetate

Dodavatel: Apollo Scientific
Nebezpečí

OR8905-1G OR8905-5G OR8905-25G
APOSOR8905-1GEA 5390 CZK
APOSOR8905-1G APOSOR8905-5G APOSOR8905-25G
3-(Bromomethyl)phenyl acetate
3-(Bromomethyl)phenyl acetate
Vzorec: C₉H₉BrO₂
Storage Temperature: Freezer
MDL Number: MFCD08543447
CAS číslo: 49617-80-3
UN: 3265
ADR: 8,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR