Tisk…

4-(2-Sulphanylethyl)pyridine

Dodavatel: Apollo Scientific
Varování

Synonyms: 2-(Pyridin-4-yl)ethyl mercaptan, 2-(Pyridin-4-yl)ethane-1-thiol, 4-(2-Mercaptoethyl)pyridine

OR8898-2.5G OR8898-500MG
APOSOR8898-2.5GEA 14490 CZK
APOSOR8898-2.5G APOSOR8898-500MG
4-(2-Sulphanylethyl)pyridine
4-(2-Sulphanylethyl)pyridine
Vzorec: C₇H₉NS
MW: 139,218 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00973403
CAS číslo: 2127-05-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR