5-Nitronicotinic acid
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
5-Nitropyridin-3-ylcarboxylic acid, 5-Nitropyridine-3-carboxylic acid, 3-Carboxy-5-nitropyridine
OR8897-1G
OR8897-5G
APOSOR8897-1GEA
2780
CZK
APOSOR8897-1G
APOSOR8897-5G
5-Nitronicotinic acid
5-Nitronicotinic acid
Vzorec:
C₆H₄N₂O₄ MW: 168,107 g/mol Bod tání: 171…173 °C Storage Temperature: Ambient |
MDL Number:
MFCD04038690 CAS číslo: 2047-49-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...