5-Methoxypyridine-2-carbonitrile
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
5-Methoxypicolinonitrile, 2-Cyano-5-methoxypyridine
OR8848-1G
OR8848-250MG
OR8848-5G
APOSOR8848-1GEA
3810
CZK
APOSOR8848-1G
APOSOR8848-250MG
APOSOR8848-5G
5-Methoxypyridine-2-carbonitrile
5-Methoxypyridine-2-carbonitrile
Vzorec:
C₇H₆N₂O MW: 134,135 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD08543467 CAS číslo: 89809-63-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...