Tisk…

4-Nitro-D-phenylalanine hydrate

Dodavatel: Apollo Scientific

OR8781-100G OR8781-25G OR8781-5G
APOSOR8781-100GEA 2360 CZK
APOSOR8781-100G APOSOR8781-25G APOSOR8781-5G
4-Nitro-D-phenylalanine hydrate
4-Nitro-D-phenylalanine hydrate
Vzorec: C₉H₁₀N₂O₄·H₂O
Storage Temperature: Refrigerator
MDL Number: MFCD00150962
CAS číslo: 444777-67-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR