4-(4-Methylhomopiperazin-1-yl)benzylamine 97%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
4-(4-Methyl-1,4-diazepan-1-yl)benzylamine
OR8689-1G
OR8689-250MG
APOSOR8689-1GEA
9200
CZK
APOSOR8689-1G
APOSOR8689-250MG
4-(4-Methylhomopiperazin-1-yl)benzylamine 97%
4-(4-Methylhomopiperazin-1-yl)benzylamine
Vzorec:
C₁₃H₂₁N₃ MW: 219,326 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08690316 CAS číslo: 448934-01-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...