Tisk…

4-(Aminomethyl)-1-methyl-1H-indole 97%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 1-(1-Methyl-1H-indol-4-yl)methylamine

OR8682-1G OR8682-250MG
APOSOR8682-1GEA 9920 CZK
APOSOR8682-1G APOSOR8682-250MG
4-(Aminomethyl)-1-methyl-1H-indole 97%
4-(Aminomethyl)-1-methyl-1H-indole
Vzorec: C₁₀H₁₂N₂
MW: 160,216 g/mol
Bod tání: 43…47 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08690264
CAS číslo: 864264-02-8
UN: 3259
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR