Tisk…

6-(Aminomethyl)-1-methyl-1H-indole

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: (1-Methyl-1H-indol-6-yl)methylamine

OR8681-100MG OR8681-250MG OR8681-1G
APOSOR8681-100MGEA 2650 CZK
APOSOR8681-100MG APOSOR8681-250MG APOSOR8681-1G
6-(Aminomethyl)-1-methyl-1H-indole
6-(Aminomethyl)-1-methyl-1H-indole
Vzorec: C₁₀H₁₂N₂
MW: 160.216 g/mol
Bod tání: 46…48 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08690253
CAS číslo: 864264-03-9
UN: 3259
ADR: 8 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR