Tisk…

4-(4-Methylhomopiperazin-1-yl)benzaldehyde 97%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 1-(4-Formylphenyl)-4-methylhomopiperazine, 4-(4-Methyl-1,4-diazepan-1-yl)benzaldehyde

OR8679-1G OR8679-5G
APOSOR8679-1GEA 20500 CZK
APOSOR8679-1G APOSOR8679-5G
4-(4-Methylhomopiperazin-1-yl)benzaldehyde 97%
4-(4-Methylhomopiperazin-1-yl)benzaldehyde
Vzorec: C₁₃H₁₈N₂O
MW: 218,295 g/mol
Bod tání: 44…46 °C
Storage Temperature: Ambient
MDL Number: MFCD08690314
CAS číslo: 166438-86-4
UN: 3259
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR