Tisk…

3-Amino-5-(methoxycarbonyl)benzeneboronic acid, pinacol ester 95%

Dodavatel: Apollo Scientific

Synonyms: 3-Amino-5-methoxycarbonylphenylboronic acid, pinacol ester, Methyl 3-amino-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate, Methyl 3-Amino-5-boronobenzoate, pinacol ester

OR8576-1G OR8576-250MG
APOSOR8576-1GEA 5720 CZK
APOSOR8576-1G APOSOR8576-250MG
3-Amino-5-(methoxycarbonyl)benzeneboronic acid, pinacol ester 95%
3-Amino-5-(methoxycarbonyl)benzeneboronic acid, pinacol ester
Vzorec: C₁₄H₂₀BNO₄
MW: 277.124 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09258776
CAS číslo: 850689-27-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR