4-(3-Bromopropoxy)benzeneboronic acid, pinacol ester 98%
Dodavatel: Apollo Scientific
Synonyms:
2-[4-(3-Bromopropoxy)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-(4-(3-Bromopropoxy)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
OR8572-1G
OR8572-5G
APOSOR8572-1GEA
2860
CZK
APOSOR8572-1G
APOSOR8572-5G
4-(3-Bromopropoxy)benzeneboronic acid, pinacol ester 98%
4-(3-Bromopropoxy)benzeneboronic acid, pinacol ester
Vzorec:
C₁₅H₂₂BBrO₃ MW: 341,048 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09258657 CAS číslo: 957061-13-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...