4-Bromothiophene-3-boronic acid 97%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
(4-Bromothiophen-3-yl)boronic acid, 3-Bromothiophene-4-boronic acid
OR8555-1G
OR8555-250MG
APOSOR8555-1GEA
1370
CZK
APOSOR8555-1G
APOSOR8555-250MG
4-Bromothiophene-3-boronic acid 97%
4-Bromothiophene-3-boronic acid
Vzorec:
C₄H₄BBrO₂S MW: 206.853 g/mol Bod tání: 196…198 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD06801663 CAS číslo: 101084-76-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...