5-Bromo-2-(piperazin-1-yl)pyridine
Dodavatel: Apollo Scientific
|
Nebezpečí
|
OR8544-25G
OR8544-5G
APOSOR8544-25GEA
10190
CZK
APOSOR8544-25G
APOSOR8544-5G
5-Bromo-2-(piperazin-1-yl)pyridine
5-Bromo-2-(piperazin-1-yl)pyridine
Vzorec:
C₉H₁₂BrN₃ MW: 242,116 g/mol Bod tání: 72 °C Storage Temperature: Ambient |
MDL Number:
MFCD04115056 CAS číslo: 73406-97-0 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...