Tisk…

4-(3-Bromophenyl)piperazine, N1-BOC protected

Dodavatel: Apollo Scientific
Varování

Synonyms: 1-Bromo-3-[4-(tert-butoxycarbonyl)piperazin-1-yl]benzene, 4-(3-Bromophenyl)-1-(tert-butoxycarbonyl)piperazine, tert-Butyl 4-(3-bromophenyl)piperazine-1-carboxylate

OR8540-1G OR8540-5G
APOSOR8540-1GEA 2570 CZK
APOSOR8540-1G APOSOR8540-5G
4-(3-Bromophenyl)piperazine, N1-BOC protected
4-(3-Bromophenyl)piperazine, N1-BOC protected
Vzorec: C₁₅H₂₁BrN₂O₂
MW: 341.243 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07781211
CAS číslo: 327030-39-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR