Tisk…

Ethyl (4-bromophenoxy)acetate

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: (4-Bromophenoxy)acetic acid ethyl ester, Ethyl 2-(4-bromophenoxy)ethanoate

OR8536-5G OR8536-250MG OR8536-1G
APOSOR8536-5GEA 0 CZK
APOSOR8536-5G APOSOR8536-250MG APOSOR8536-1G
Ethyl (4-bromophenoxy)acetate
Ethyl (4-bromophenoxy)acetate
Vzorec: C₁₀H₁₁BrO₃
MW: 259,097 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00462746
CAS číslo: 6964-29-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR