Tisk…

3-(2,2-Dicyanovinyl)benzeneboronic acid 98%

Dodavatel: Apollo Scientific

OR8451-1G OR8451-5G
APOSOR8451-1GEA 5890 CZK
APOSOR8451-1G APOSOR8451-5G
3-(2,2-Dicyanovinyl)benzeneboronic acid 98%
3-(2,2-Dicyanovinyl)benzeneboronic acid
Vzorec: C₁₀H₇BN₂O₂
Storage Temperature: Ambient
MDL Number: MFCD06214067
CAS číslo: 957034-31-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR