Tisk…

1H-Imidazole-1-carbonitrile 94%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 1-Cyano-1H-imidazole

OR8175T-1G OR8175T-250MG OR8175T-5G
APOSOR8175T-1GEA 6620 CZK
APOSOR8175T-1G APOSOR8175T-250MG APOSOR8175T-5G
1H-Imidazole-1-carbonitrile 94%
1H-Imidazole-1-carbonitrile
Vzorec: C₄H₃N₃
MW: 93,087 g/mol
Bod varu: 185…189 °C
Bod tání: 59,5…60.5 °C
Storage Temperature: Freezer
MDL Number: MFCD00269955
CAS číslo: 36289-36-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR