Tisk…

Benzyl-2-acetamido-2-deoxy-α-D-galactopyranoside 98%

Dodavatel: Apollo Scientific

OR8162-100MG OR8162-500MG
APOSOR8162-100MGEA 9130 CZK
APOSOR8162-100MG APOSOR8162-500MG
Benzyl-2-acetamido-2-deoxy-α-D-galactopyranoside 98%
Benzyl-2-acetamido-2-deoxy-α-D-galactopyranoside
Vzorec: C₁₅H₂₁NO₆
MW: 311,33 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00057920
CAS číslo: 3554-93-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR