Tisk…

1-(4-Methylthiophenyl)-2-methyl-5-phenylpyrrole-3-carboxylic acid 95+%

Dodavatel: Apollo Scientific

OR8137-10G OR8137-1G OR8137-5G
APOSOR8137-10GEA 24600 CZK
APOSOR8137-10G APOSOR8137-1G APOSOR8137-5G
1-(4-Methylthiophenyl)-2-methyl-5-phenylpyrrole-3-carboxylic acid 95+%
1-(4-Methylthiophenyl)-2-methyl-5-phenylpyrrole-3-carboxylic acid
Vzorec: C₁₉H₁₇NO₂S
MW: 323,409 g/mol
Bod tání: 202…203 °C
Storage Temperature: Ambient
MDL Number: MFCD02180787
CAS číslo: 306936-46-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR