Tisk…

3-(2-Formyl-1H-pyrrol-1-yl)propanenitrile 97%

Dodavatel: Apollo Scientific
Varování

Synonyms: 1-(2-Cyanoethyl)-2-formyl-1H-pyrrole, 1-(2-Cyanoethyl)-1H-pyrrole-2-carboxaldehyde

OR8068-10G OR8068-1G
APOSOR8068-10GEA 9760 CZK
APOSOR8068-10G APOSOR8068-1G
3-(2-Formyl-1H-pyrrol-1-yl)propanenitrile 97%
3-(2-Formyl-1H-pyrrol-1-yl)propanenitrile
Vzorec: C₈H₈N₂O
MW: 148,162 g/mol
Bod varu: 130…132 °C (1,0 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD02180764
CAS číslo: 43036-05-1
UN: 3276
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR