6-[4-(tert-Butyl)phenoxy]pyridin-3-amine 97%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
5-Amino-2-[4-(tert-butyl)phenoxy]pyridine, CB-103, 6-(4-(tert-Butyl)phenoxy)pyridin-3-amine
OR8051-1G
OR8051-5G
OR8051-10G
APOSOR8051-1GEA
2400
CZK
APOSOR8051-1G
APOSOR8051-5G
APOSOR8051-10G
6-[4-(tert-Butyl)phenoxy]pyridin-3-amine 97%
6-[4-(tert-Butyl)phenoxy]pyridin-3-amine
Vzorec:
C₁₅H₁₈N₂O MW: 242,316 g/mol Bod tání: 89…90 °C Storage Temperature: Ambient |
MDL Number:
MFCD00052647 CAS číslo: 218457-67-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...