Tisk…

5-Amino-1H-indole-3-carboxamide hydrochloride, tech

Dodavatel: Apollo Scientific
Varování

Synonyms: 3-(Aminocarbonyl)-1H-indol-5-amine hydrochloride

OR8008-1G OR8008-250MG
APOSOR8008-1GEA 4900 CZK
APOSOR8008-1G APOSOR8008-250MG
5-Amino-1H-indole-3-carboxamide hydrochloride, tech
5-Amino-1H-indole-3-carboxamide hydrochloride
Vzorec: C₉H₉N₃O·ClH
Bod tání: 296…298 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02259416
CAS číslo: 306936-36-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR