4-(5-Acetylthien-2-yl)-2-methylbut-3-yn-2-ol 98%
Dodavatel: Apollo Scientific
OR7854-1G
OR7854-5G
APOSOR7854-1GEA
5200
CZK
APOSOR7854-1G
APOSOR7854-5G
4-(5-Acetylthien-2-yl)-2-methylbut-3-yn-2-ol 98%
4-(5-Acetylthien-2-yl)-2-methylbut-3-yn-2-ol
Vzorec:
C₁₁H₁₂O₂S MW: 208,277 g/mol Bod tání: 79…82 °C Storage Temperature: Ambient |
MDL Number:
MFCD00173926 CAS číslo: 175203-51-7 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...