Tisk…

5-Bromo-4-methylthiophene-2-carboxaldehyde

Dodavatel: Apollo Scientific
Varování

OR7818-100G OR7818-25G OR7818-5G
APOSOR7818-100GEA 15050 CZK
APOSOR7818-100G APOSOR7818-25G APOSOR7818-5G
5-Bromo-4-methylthiophene-2-carboxaldehyde
5-Bromo-4-methylthiophene-2-carboxaldehyde
Vzorec: C₆H₅BrOS
Storage Temperature: Refrigerator
MDL Number: MFCD06657807
CAS číslo: 189331-47-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR