Tisk…

1-(2,6-Dimethoxybenzyl)piperidin-4-amine dihydrochloride

Dodavatel: Apollo Scientific
Varování

Synonyms: 4-Amino-1-(2,6-dimethoxybenzyl)piperidine dihydrochloride, 1-(2,6-Dimethoxybenzyl)piperidin-4-ylaminedihydrochloride, 1-[(2,6-Dimethoxyphenyl)methyl]piperidin-4-amine dihydrochloride

OR7723-1G OR7723-250MG
APOSOR7723-1GEA 2070 CZK
APOSOR7723-1G APOSOR7723-250MG
1-(2,6-Dimethoxybenzyl)piperidin-4-amine dihydrochloride
1-(2,6-Dimethoxybenzyl)piperidin-4-amine dihydrochloride
Vzorec: C₁₄H₂₂N₂O₂·2HCl
MW: 323,26 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06247029
CAS číslo: 1185303-17-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR