Methyl 4-ethylbenzoate 98%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
4-Ethylbenzoic acid methyl ester
OR7700-1G
OR7700-5G
APOSOR7700-1GEA
1330
CZK
APOSOR7700-1G
APOSOR7700-5G
Methyl 4-ethylbenzoate 98%
Methyl 4-ethylbenzoate
Vzorec:
C₁₀H₁₂O₂ MW: 164,201 g/mol Bod varu: 105…107 °C (15 mm Hg) Storage Temperature: Ambient |
MDL Number:
MFCD00221477 CAS číslo: 7364-20-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...