Tisk…

1H-Benzimidazole-2-carboxylic acid hydrate 90+%

Dodavatel: Apollo Scientific
Varování

Synonyms: 2-Carboxy-1H-benzimidazole hydrate

OR7662-1G OR7662-250MG
APOSOR7662-1GEA 1990 CZK
APOSOR7662-1G APOSOR7662-250MG
1H-Benzimidazole-2-carboxylic acid hydrate 90+%
1H-Benzimidazole-2-carboxylic acid hydrate
Vzorec: C₈H₆N₂O₂·H₂O
MW: 180,161 g/mol
Bod tání: 169…171 °C
Storage Temperature: Ambient
MDL Number: MFCD07368500
CAS číslo: 849776-47-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR