2-Chloro-4-(methylsulphonyl)benzaldehyde hydrate 95+%
Dodavatel: Apollo Scientific
|
Varování
|
OR7624-1G
OR7624-5G
APOSOR7624-1GEA
7540
CZK
APOSOR7624-1G
APOSOR7624-5G
2-Chloro-4-(methylsulphonyl)benzaldehyde hydrate 95+%
2-Chloro-4-(methylsulphonyl)benzaldehyde hydrate
Vzorec:
C₈H₇ClO₃S·H₂O MW: 236,673 g/mol Bod tání: 111…112 °C (rozkl.) Storage Temperature: Ambient |
MDL Number:
MFCD08166750 CAS číslo: 1171826-50-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...