Tisk…

5-(2-Methylphenyl)-2-furoic acid

Dodavatel: Apollo Scientific
Varování

OR7574-1G OR7574-5G
APOSOR7574-1GEA 20470 CZK
APOSOR7574-1G APOSOR7574-5G
5-(2-Methylphenyl)-2-furoic acid
5-(2-Methylphenyl)-2-furoic acid
Vzorec: C₁₂H₁₀O₃
MW: 202.206 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04039143
CAS číslo: 80174-04-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR