Tisk…

4-(5-Methylfur-2-yl)benzaldehyde

Dodavatel: Apollo Scientific
Varování

Synonyms: 2-(4-Formylphenyl)-5-methylfuran

OR7542-1G OR7542-250MG
APOSOR7542-1GEA 4960 CZK
APOSOR7542-1G APOSOR7542-250MG
4-(5-Methylfur-2-yl)benzaldehyde
4-(5-Methylfur-2-yl)benzaldehyde
Vzorec: C₁₂H₁₀O₂
MW: 186,207 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04039125
CAS číslo: 400748-10-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR