Tisk…

2-Amino-3'-methylbiphenyl hydrochloride

Dodavatel: Apollo Scientific
Varování

Synonyms: 2-(3-Methylphenyl)aniline hydrochloride, 3'-Methyl-[1,1'-biphenyl]-2-amine hydrochloride

OR7491-100MG OR7491-250MG OR7491-1G
APOSOR7491-100MGEA 6950 CZK
APOSOR7491-100MG APOSOR7491-250MG APOSOR7491-1G
2-Amino-3'-methylbiphenyl hydrochloride
2-Amino-3'-methylbiphenyl hydrochloride
Vzorec: C₁₃H₁₃N·ClH
MW: 219,71 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06739418
CAS číslo: 139769-13-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR