Tisk…

2-Hydroxy-5-(phenylethynyl)benzoic acid

Dodavatel: Apollo Scientific
Varování

Synonyms: 2-Carboxy-4-(phenylethynyl)phenol, 5-(Phenylethynyl)salicylic acid

OR7432-1G OR7432-250MG OR7432-50MG
APOSOR7432-1GEA 9790 CZK
APOSOR7432-1G APOSOR7432-250MG APOSOR7432-50MG
2-Hydroxy-5-(phenylethynyl)benzoic acid
2-Hydroxy-5-(phenylethynyl)benzoic acid
Vzorec: C₁₅H₁₀O₃
MW: 238,238 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04039086
CAS číslo: 32192-74-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR