3-(Fur-2-yl)benzaldehyde
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
2-(3-Formylphenyl)furan
OR7397-1G
OR7397-250MG
APOSOR7397-1GEA
5200
CZK
APOSOR7397-1G
APOSOR7397-250MG
3-(Fur-2-yl)benzaldehyde
3-(Fur-2-yl)benzaldehyde
Vzorec:
C₁₁H₈O₂ MW: 172,18 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03990461 CAS číslo: 85553-52-2 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...