Tisk…

3-(Fur-2-yl)aniline hydrochloride

Dodavatel: Apollo Scientific
Varování

Synonyms: 2-(3-Aminophenyl)furan hydrochloride

OR7393-1G OR7393-5G
APOSOR7393-1GEA 3640 CZK
APOSOR7393-1G APOSOR7393-5G
3-(Fur-2-yl)aniline hydrochloride
3-(Fur-2-yl)aniline hydrochloride
Vzorec: C₁₀H₉NO·ClH
MW: 195,645 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07366478
CAS číslo: 1172481-22-9
UN: 2811
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR