Tisk…

3-(3-Chlorophenyl)prop-2-yn-1-ol

Dodavatel: Apollo Scientific
Varování

OR7381-1G OR7381-2.5G OR7381-250MG
APOSOR7381-1GEA 3110 CZK
APOSOR7381-1G APOSOR7381-2.5G APOSOR7381-250MG
3-(3-Chlorophenyl)prop-2-yn-1-ol
3-(3-Chlorophenyl)prop-2-yn-1-ol
Vzorec: C₉H₇ClO
MW: 166,604 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04039064
CAS číslo: 80151-33-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR