Tisk…

4-[(6-Chloro-2,3-dihydro-3-oxo-4H-1,4-benzoxazin-4-yl)methyl]benzoic acid

Dodavatel: Apollo Scientific
Varování

Synonyms: 4-[(6-Chloro-2,3-dihydro-3-oxobenzo[b][1,4]oxazin-4-yl)methyl]benzoic acid, 4-(4-Carboxybenzyl)-6-chloro-2,3-dihydro-3-oxo-4H-1,4-benzoxazine

OR7372-10MG OR7372-50MG
APOSOR7372-10MGEA 622 CZK
APOSOR7372-10MG APOSOR7372-50MG
4-[(6-Chloro-2,3-dihydro-3-oxo-4H-1,4-benzoxazin-4-yl)methyl]benzoic acid
4-[(6-Chloro-2,3-dihydro-3-oxo-4H-1,4-benzoxazin-4-yl)methyl]benzoic acid
Vzorec: C₁₆H₁₂ClNO₄
MW: 317,724 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00269589
CAS číslo: 874605-63-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR