4'-Chloro-[1,1'-biphenyl]-2-carboxaldehyde
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
4'-Chloro-1,1'-biphenyl-2-carboxaldehyde, 2-(4-Chlorophenyl)benzaldehyde, 4'-Chloro-2-formylbiphenyl
OR7355-1G
OR7355-500MG
APOSOR7355-1GEA
12830
CZK
APOSOR7355-1G
APOSOR7355-500MG
4'-Chloro-[1,1'-biphenyl]-2-carboxaldehyde
4'-Chloro-[1,1'-biphenyl]-2-carboxaldehyde
Vzorec:
C₁₃H₉ClO MW: 216,663 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03424629 CAS číslo: 153850-83-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...