1-O-Acetyl-2-deoxy-3,5-di-O-benzoylribofuranose
Dodavatel: Apollo Scientific
|
Varování
|
OR7350T-250MG
OR7350T-50MG
APOSOR7350T-250MGEA
24630
CZK
APOSOR7350T-250MG
APOSOR7350T-50MG
1-O-Acetyl-2-deoxy-3,5-di-O-benzoylribofuranose
1-O-Acetyl-2-deoxy-3,5-di-O-benzoylribofuranose
Vzorec:
C₂₁H₂₀O₇ MW: 384.379 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00269868 CAS číslo: 51255-12-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...