Tisk…

3'-Chloro-[1,1'-biphenyl]-3-carboxaldehyde

Dodavatel: Apollo Scientific
Varování

OR7348-1G OR7348-500MG
APOSOR7348-1GEA 13860 CZK
APOSOR7348-1G APOSOR7348-500MG
3'-Chloro-[1,1'-biphenyl]-3-carboxaldehyde
3'-Chloro-[1,1'-biphenyl]-3-carboxaldehyde
Vzorec: C₁₃H₉ClO
MW: 216.663 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03424621
CAS číslo: 400745-60-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR