Tisk…

Acetone 4-bromo-2-nitrophenylhydrazone

Dodavatel: Apollo Scientific
Varování

OR7328-1G OR7328-500MG
APOSOR7328-1GEA 2900 CZK
APOSOR7328-1G APOSOR7328-500MG
Acetone 4-bromo-2-nitrophenylhydrazone
Acetone 4-bromo-2-nitrophenylhydrazone
Vzorec: C₉H₈BrN₃O₃
MW: 286,082 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08445499
CAS číslo: 914636-18-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR