Tisk…

4-(n-Octyl)benzeneboronic acid 97%

Dodavatel: Apollo Scientific
Varování

Synonyms: (4-Octylphenyl)boronic acid

OR7246-5G OR7246-1G OR7246-250MG
APOSOR7246-5GEA 29300 CZK
APOSOR7246-5G APOSOR7246-1G APOSOR7246-250MG
4-(n-Octyl)benzeneboronic acid 97%
4-(n-Octyl)benzeneboronic acid
Vzorec: C₁₄H₂₃BO₂
MW: 234,142 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04039034
CAS číslo: 133997-05-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR