4-Benzyloxy-3-formylbenzeneboronic acid
Dodavatel: Apollo Scientific
Synonyms:
3-Formyl-4-benzyloxyphenylboronic acid, (4-(Benzyloxy)-3-formylphenyl)boronic acid
OR7159-1G
OR7159-5G
APOSOR7159-1GEA
3940
CZK
APOSOR7159-1G
APOSOR7159-5G
4-Benzyloxy-3-formylbenzeneboronic acid
4-Benzyloxy-3-formylbenzeneboronic acid
Vzorec:
C₁₄H₁₃BO₄ MW: 256,062 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD02179487 CAS číslo: 121124-98-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...