Tisk…

Ethyl 6-bromo-2-chloro-3-ethoxybenzoate 97%

Dodavatel: Apollo Scientific

OR71011-1G OR71011-500MG OR71011-5G
APOSOR71011-1GEA 6550 CZK
APOSOR71011-1G APOSOR71011-500MG APOSOR71011-5G
Ethyl 6-bromo-2-chloro-3-ethoxybenzoate 97%
Ethyl 6-bromo-2-chloro-3-ethoxybenzoate
Vzorec: C₁₁H₁₂BrClO₃
Storage Temperature: Refrigerator
MDL Number: MFCD32874088
CAS číslo: 2384335-29-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR