Tisk…

Methyl 6-bromo-2-chloro-3-ethoxybenzoate 97%

Dodavatel: Apollo Scientific

OR71009-1G OR71009-500MG
APOSOR71009-1GEA 6550 CZK
APOSOR71009-1G APOSOR71009-500MG
Methyl 6-bromo-2-chloro-3-ethoxybenzoate 97%
Methyl 6-bromo-2-chloro-3-ethoxybenzoate
Vzorec: C₁₀H₁₀BrClO₃
MW: 293,542 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD32874090
CAS číslo: 2384926-02-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR