Tisk…

Tributylsulphoammonium betaine 95%

Dodavatel: Apollo Scientific

Synonyms: 1-[Dibutyl(sulphonato)azaniumyl]butane

OR71000-2G OR71000-500MG
APOSOR71000-2GEA 12240 CZK
APOSOR71000-2G APOSOR71000-500MG
Tributylsulphoammonium betaine 95%
Tributylsulphoammonium betaine
Vzorec: C₁₂H₂₇NO₃S
MW: 265.413 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32672293
CAS číslo: 2364603-74-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR