4,4'-Bipyridine dihydrochloride
Dodavatel: Apollo Scientific
|
Nebezpečí
|
OR7090-1G
OR7090-250MG
OR7090-5G
APOSOR7090-1GEA
3940
CZK
APOSOR7090-1G
APOSOR7090-250MG
APOSOR7090-5G
4,4'-Bipyridine dihydrochloride
4,4'-Bipyridine dihydrochloride
Vzorec:
C₁₀H₈N₂·2ClH Bod tání: 302…306 °C Hustota: 1,52 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00012824 CAS číslo: 27926-72-3 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...