Tisk…

3,5-Dimethyl-4-[2-(methylamino)ethyl]-1H-pyrazole dihydrochloride

Dodavatel: Apollo Scientific
Varování

OR7084-1G OR7084-250MG
APOSOR7084-1GEA 4960 CZK
APOSOR7084-1G APOSOR7084-250MG
3,5-Dimethyl-4-[2-(methylamino)ethyl]-1H-pyrazole dihydrochloride
3,5-Dimethyl-4-[2-(methylamino)ethyl]-1H-pyrazole dihydrochloride
Vzorec: C₈H₁₅N₃·2ClH
Storage Temperature: Ambient
MDL Number: MFCD07366477
CAS číslo: 423176-42-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR