2-Amino-5-chloro-3-iodobenzoic acid 95%
Dodavatel: Apollo Scientific
|
Varování
|
OR70216-1G
OR70216-5G
OR70216-250MG
APOSOR70216-1GEA
705
CZK
APOSOR70216-1G
APOSOR70216-5G
APOSOR70216-250MG
2-Amino-5-chloro-3-iodobenzoic acid 95%
2-Amino-5-chloro-3-iodobenzoic acid
Vzorec:
C₇H₅ClINO₂ Bod tání: 246…247 °C Storage Temperature: Ambient |
MDL Number:
MFCD07780627 CAS číslo: 64724-23-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...