Bicyclo[3.1.0]hexan-2-one 97%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
2-Oxobicyclo[3.1.0]hexane
OR70212-1G
OR70212-250MG
APOSOR70212-1GEA
8070
CZK
APOSOR70212-1G
APOSOR70212-250MG
Bicyclo[3.1.0]hexan-2-one 97%
Bicyclo[3.1.0]hexan-2-one
Vzorec:
C₆H₈O MW: 96,127 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD20040018 CAS číslo: 4160-49-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...